Learn More
Medchemexpress LLC Cis/trans-nepetalactol | 109215-55-6 | 95.0% | 168.23 g/mol | C10H16O2 | 5 MG
SDP
Catalog No. 5000285117
Encompass_Preferred
Cis/trans-nepetalactol is an iridoid natural product found in Actinidia polygama (silver vine). It is reported to be a major cat attractant and to exhibit mosquito-repellent activity. Supplied for research use, the compound is typically provided as a purified solid useful for behavioral, ecological, and chemical studies.
  • Natural iridoid compound from silver vine.
  • Acts as a cat attractant and insect repellent.
  • Provided as a purified research-grade material (95.0% purity).
  • Available in small milligram pack sizes suitable for assays.
  • Useful as a reference standard or reagent in behavioral and chemical ecology studies.
Catalog No. 50-002-85117 Supplier Medchemexpress LLC Supplier No. HYW5874155MG
Cis/trans-nepetalactol is an iridoid natural product found in Actinidia polygama (silver vine). It is reported to be a major cat attractant and to exhibit mosquito-repellent activity. Supplied for research use, the compound is typically provided as a purified solid useful for behavioral, ecological, and chemical studies.
  • Natural iridoid compound from silver vine.
  • Acts as a cat attractant and insect repellent.
  • Provided as a purified research-grade material (95.0% purity).
  • Available in small milligram pack sizes suitable for assays.
  • Useful as a reference standard or reagent in behavioral and chemical ecology studies.