Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cis/trans-nepetalactol | 109215-55-6 | MFCD31692437 | 95.0% | 168.23 | C10H16O2 | 10 MG
SDP

Catalog No. 5000285077
Encompass_Preferred

cis/trans-Nepetalactol is an iridoid compound found in silver vine (Actinidia polygama). It is reported to act as a cat attractant and to exhibit mosquito-repellent activity. The material is intended for research use and is supplied with a certificate of analysis documenting purity and molecular data.

  • Natural iridoid found in Actinidia polygama.
  • Acts as a cat attractant and mosquito repellent.
  • Useful for behavioral and entomology research.
  • Supplied with certificate of analysis verifying purity.
  • Purity typically around 95.0% (NMR).
  • Appearance: colorless to light yellow, solid-liquid mixture.
  • Recommended storage: -20°C for pure material, and lower temperatures for solutions as specified.

Catalog No. 50-002-85077 Supplier Medchemexpress LLC Supplier No. HYW58741510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cis/trans-Nepetalactol is an iridoid compound found in silver vine (Actinidia polygama). It is reported to act as a cat attractant and to exhibit mosquito-repellent activity. The material is intended for research use and is supplied with a certificate of analysis documenting purity and molecular data.

  • Natural iridoid found in Actinidia polygama.
  • Acts as a cat attractant and mosquito repellent.
  • Useful for behavioral and entomology research.
  • Supplied with certificate of analysis verifying purity.
  • Purity typically around 95.0% (NMR).
  • Appearance: colorless to light yellow, solid-liquid mixture.
  • Recommended storage: -20°C for pure material, and lower temperatures for solutions as specified.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.