Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis,trans,trans-1,5,9-Cyclododecatriene, TCI America™
SDP

Catalog No. C0463500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0463500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0463500ML Supplier TCI America Supplier No. C0463500ML

missing translation for 'validMainframeMsg'

Specifications

CAS 706-31-0
Color Colorless
Boiling Point 242°C
Molecular Formula C12H18
Quantity 500 mL
MDL Number MFCD00063609
UN Number 2518
Synonym unii-7k92xvu2c2, trans,trans,cis-1,5,9-cyclododecatriene, 1e,5e,9z-cyclododeca-1,5,9-triene, 1,5,9-cyclododecatriene, 1e,5e,9z, cis,trans,trans-1,5,9-cyclododecatriene, cyclododecatriene-1,5,9, cyclododeca-1,5,9-triene, 1,5,9-cyclododecatriene, e,e,z-1,5,9-cyclododecatriene, trans-trans-cis-1,5,9-cyclodecatriene
InChI Key ZOLLIQAKMYWTBR-RYMQXAEESA-N
SMILES C1CC=CCCC=CCCC=C1
IUPAC Name (1Z,5E,9E)-cyclododeca-1,5,9-triene
Molecular Weight (g/mol) 162.276
PubChem CID 6262774
Formula Weight 162.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material cis,trans,trans-1,5,9-Cyclododecatriene
Show More Show Less
EINECSNumber (3)-2239
RTECSNumber GU2310000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.