Learn More
TARGETMOL CHEMICALS INC CIS-TRISMETHOXY RESVERAT 10MG
SDP
Catalog No. 502576961
Encompass_Preferred
Also available in 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. cis-trismethoxy Resveratrol ((Z)-354'-Trimethoxystilbene) inhibits tubulin polymerization (IC50 = 4 μM) and has anti-mitotic effects. purity: 97%
Catalog No. 50-257-6961 Supplier TARGETMOL CHEMICALS INC Supplier No. T3330L10MG
Also available in 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. cis-trismethoxy Resveratrol ((Z)-354'-Trimethoxystilbene) inhibits tubulin polymerization (IC50 = 4 μM) and has anti-mitotic effects. purity: 97%