Learn More
Medchemexpress LLC HY-113008A 10mg Medchemexpress, cis-Urocanic acid CAS:7699-35-6 Purity:>98%
SDP
Catalog No. 501969214
Encompass_Preferred
Medchemexpress, HY-113008A 10mg cis-Urocanic acid CAS:7699-35-6 cis-Urocanic acid is a 5-HT2A receptor agonist. cis-Urocanic acid binds to 5-HT receptor with relatively high affinity (Kd=4.6 nM). cis-Urocanic acid is an immune modulator that induces immunosuppression by binding to the 5-HT2A receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9214 Supplier Medchemexpress LLC Supplier No. HY113008A10MG
Medchemexpress, HY-113008A 10mg cis-Urocanic acid CAS:7699-35-6 cis-Urocanic acid is a 5-HT2A receptor agonist. cis-Urocanic acid binds to 5-HT receptor with relatively high affinity (Kd=4.6 nM). cis-Urocanic acid is an immune modulator that induces immunosuppression by binding to the 5-HT2A receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.