Learn More
Medchemexpress LLC Cis-urocanic acid-13c3 | 7699-35-6 | 99.5% | 139.09 | 1 MG
SDP
Catalog No. 5000439758
Encompass_Preferred
cis-Urocanic acid-13C3 is the 13C-labeled cis-Urocanic acid. cis-Urocanic acid is a 5-HT2A receptor agonist and an immune modulator that induces immunosuppression by binding to the 5-HT2A receptor with a relatively high affinity (Kd=4.6 nM).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
Catalog No. 50-004-39758 Supplier Medchemexpress LLC Supplier No. HY113008AS1MG
Add to Cart
Add to Cart
cis-Urocanic acid-13C3 is the 13C-labeled cis-Urocanic acid. cis-Urocanic acid is a 5-HT2A receptor agonist and an immune modulator that induces immunosuppression by binding to the 5-HT2A receptor with a relatively high affinity (Kd=4.6 nM).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.