Learn More
TARGETMOL CHEMICALS INC CIS-UROCANIC ACID 5MG
SDP
Catalog No. 502581654
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. cis-Urocanic acid ((E)-Urocanic acid) is a 5-HT2A receptor agonist (Kd 4.6 nM). It is an immune modulator that induces immunosuppression by binding to the 5-HT2A receptor. purity: 99%
Catalog No. 50-258-1654 Supplier TARGETMOL CHEMICALS INC Supplier No. T108245MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. cis-Urocanic acid ((E)-Urocanic acid) is a 5-HT2A receptor agonist (Kd 4.6 nM). It is an immune modulator that induces immunosuppression by binding to the 5-HT2A receptor. purity: 99%