Learn More
Cayman Chemical cIs-VaccenIc AcId 100mg
SDP
Catalog No. 502983255
Encompass_Preferred
An ω-7 fatty acid; induces differentiation of, and γ-globin synthesis in, K562 and JK-1 cells, as well as isolated sickle cell TMbmEPSCs
Catalog No. 50-298-3255 Supplier Cayman Chemical Supplier No. 20023100 MG
An ω-7 fatty acid; induces differentiation of, and γ-globin synthesis in, K562 and JK-1 cells, as well as isolated sickle cell TMbmEPSCs