Learn More
Cayman Chemical cis-Vaccenic Acid, Purity: ≥98%, CAS: 506-17-2, 500 mg
SDP
Catalog No. 502984188
Encompass_Preferred
An ω-7 fatty acid; induces differentiation of, and γ-globin synthesis in, K562 and JK-1 cells, as well as isolated sickle cell TMbmEPSCs
Catalog No. 50-298-4188 Supplier Cayman Chemical Supplier No. 20023500 MG
An ω-7 fatty acid; induces differentiation of, and γ-globin synthesis in, K562 and JK-1 cells, as well as isolated sickle cell TMbmEPSCs