Learn More
Cayman Chemical cis-Vaccenic Acid, Purity: ≥98%, CAS: 506-17-2, 50 mg
SDP
Catalog No. 502984023
Encompass_Preferred
An ω-7 fatty acid; induces differentiation of, and γ-globin synthesis in, K562 and JK-1 cells, as well as isolated sickle cell TMbmEPSCs
Catalog No. 50-298-4023 Supplier Cayman Chemical Supplier No. 2002350 MG
An ω-7 fatty acid; induces differentiation of, and γ-globin synthesis in, K562 and JK-1 cells, as well as isolated sickle cell TMbmEPSCs