Learn More
Cayman Chemical cis-Vaccenic Acid-d13, Purity: ≥99% deuterated forms (d1-d13), 10 mg
SDP
Catalog No. 502941545
Encompass_Preferred
An internal standard for the quantification of cis-vaccenic acid by GC- or LC-MS
Catalog No. 50-294-1545 Supplier Cayman Chemical Supplier No. 2771610 MG
An internal standard for the quantification of cis-vaccenic acid by GC- or LC-MS