Learn More
Cayman Chemical cIs-VaccenIc AcId-d13 1mg
SDP
Catalog No. 502940272
Encompass_Preferred
An internal standard for the quantification of cis-vaccenic acid by GC- or LC-MS
Catalog No. 50-294-0272 Supplier Cayman Chemical Supplier No. 277161 MG
An internal standard for the quantification of cis-vaccenic acid by GC- or LC-MS