Learn More
Cayman Chemical cis-Vaccenic Acid-d13, Purity: ≥99% deuterated forms (d1-d13), 5 mg
SDP
Catalog No. 502937543
Encompass_Preferred
An internal standard for the quantification of cis-vaccenic acid by GC- or LC-MS
Catalog No. 50-293-7543 Supplier Cayman Chemical Supplier No. 277165 MG
An internal standard for the quantification of cis-vaccenic acid by GC- or LC-MS