Learn More
Medchemexpress LLC Cis VH-298 | 2097381-86-5 | 100.0% | 523.65 g/mol | C27H33N5O4S | 5 MG
SDP
Catalog No. 5000211802
Encompass_Preferred
Cis VH-298 is the cis geometric isomer of VH-298 that shows a significant loss of binding specificity of hydroxylated HIF-1α peptides to the von Hippel-Lindau (VHL) protein in vitro. Supplied as a high-purity research-grade small molecule, it is intended for laboratory studies of VHL-HIF interactions and to serve as a stereochemical control when comparing isomer effects.
  • High purity suitable for analytical and cell-based research.
  • Provided in research-scale quantities for experimental use.
  • Useful as a stereochemical control to compare cis/trans activity.
  • Demonstrates reduced VHL binding, aiding mechanistic studies of HIF regulation.
  • Includes datasheet with chemical properties and handling information.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-11802 Supplier Medchemexpress LLC Supplier No. HY100947A5MG
Cis VH-298 is the cis geometric isomer of VH-298 that shows a significant loss of binding specificity of hydroxylated HIF-1α peptides to the von Hippel-Lindau (VHL) protein in vitro. Supplied as a high-purity research-grade small molecule, it is intended for laboratory studies of VHL-HIF interactions and to serve as a stereochemical control when comparing isomer effects.
  • High purity suitable for analytical and cell-based research.
  • Provided in research-scale quantities for experimental use.
  • Useful as a stereochemical control to compare cis/trans activity.
  • Demonstrates reduced VHL binding, aiding mechanistic studies of HIF regulation.
  • Includes datasheet with chemical properties and handling information.
  • For research use only; not for human or diagnostic use.