Learn More
Cayman Chemical cIs10hydroxy NortrIptylIn 10mg
SDP
Supplier:  Cayman Chemical 3665910 MG
Encompass_Preferred
An active metabolite of amitriptyline and nortriptyline; formed from amitriptyline and nortriptyline by CYP2D6; inhibits norepinephrine reuptake in rat cortical slices (EC50 = 160 nM)
Catalog No. 50-290-9502