Learn More
Cayman Chemical cis-7-Hexadecenoic Acid, Purity: ≥98%, CAS: 2416-19-5, 100 mg
SDP
Catalog No. 502887927
Encompass_Preferred
A mono-unsaturated fatty acid found in cellular membranes; has only been detected in strains of the genera Nitrospira, which are nitrite oxidizing autotrophic bacteria
Catalog No. 50-288-7927 Supplier Cayman Chemical Supplier No. 10007290100 MG
A mono-unsaturated fatty acid found in cellular membranes; has only been detected in strains of the genera Nitrospira, which are nitrite oxidizing autotrophic bacteria