Learn More
Medchemexpress LLC Cisapride | 81098-60-4 | MFCD00868098 | >98.0% | 465.95 | C23H29ClFN3O4 | 5 MG
SDP
Catalog No. 5000223174
Encompass_Preferred
Cisapride is a gastroprokinetic agent and a research-standard 5-HT4 receptor agonist used in pharmacology and ion-channel studies. It is reported to block hERG channels and is supplied as a high-purity analytical standard for laboratory use (CAS 81098-60-4, formula C23H29ClFN3O4, MW 465.95).
  • High purity (>98.0%) suitable for research and analytical applications.
  • Orally active 5-HT4 receptor agonist for pharmacological assays.
  • hERG blocker with reported IC50 ≈ 9.4 nM and 5-HT4 EC50 ≈ 140 nM.
  • Available as a solid in small pack sizes for analytical testing.
  • Molecular formula C23H29ClFN3O4; molecular weight 465.95.
Catalog No. 50-002-23174 Supplier Medchemexpress LLC Supplier No. HY141495MG
Cisapride is a gastroprokinetic agent and a research-standard 5-HT4 receptor agonist used in pharmacology and ion-channel studies. It is reported to block hERG channels and is supplied as a high-purity analytical standard for laboratory use (CAS 81098-60-4, formula C23H29ClFN3O4, MW 465.95).
  • High purity (>98.0%) suitable for research and analytical applications.
  • Orally active 5-HT4 receptor agonist for pharmacological assays.
  • hERG blocker with reported IC50 ≈ 9.4 nM and 5-HT4 EC50 ≈ 140 nM.
  • Available as a solid in small pack sizes for analytical testing.
  • Molecular formula C23H29ClFN3O4; molecular weight 465.95.