Learn More
Selleck Chemical LLC Cisatracurium Besylate S2113-25mg
SDP
Supplier:  Selleck Chemical LLC S211325MG
Encompass_Preferred
Cisatracurium Besylate(51W89) is a nondepolarizing neuromuscular blocking agent antagonizing the action of acetylcholine by inhibiting neuromuscular transmission
Catalog No. 50-311-0384