Learn More
TARGETMOL CHEMICALS INC Cisatracurium besylate 50MG
SDP
Catalog No. 502876394
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Cisatracurium besylate (51W89) is a nondepolarizing skeletal muscle relaxant intended for intravenous administration. Purity 98%
Catalog No. 50-287-6394 Supplier TARGETMOL CHEMICALS INC Supplier No. T145950MG
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Cisatracurium besylate (51W89) is a nondepolarizing skeletal muscle relaxant intended for intravenous administration. Purity 98%