Learn More
Apexbio Technology LLC Cisatracurium Besylate 96946-42-8 100mg
SDP
Supplier:  Apexbio Technology LLC B1355100
Encompass_Preferred
Cisatracurium Besylate (CAS 96946-42-8) is a non-depolarizing neuromuscular blocking agent that acts by competitively inhibiting acetylcholine at nicotinic receptors on the neuromuscular junction This inhibition prevents depolarization of the muscle cell membrane resulting in skeletal muscle relaxation Cisatracurium Besylate is utilized in biomedical research to study neuromuscular transmission skeletal muscle function and mechanisms underlying muscle paralysis It is also frequently used as an adjunct in anesthesia research to model conditions requiring controlled muscle relaxation such as intubation or mechanical ventilation
Catalog No. 50-000-89790