Learn More
Aviva Systems Biology Corp CISD1 Antibody - C-terminal region (ARP65647_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65647_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CISD1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human CISD1. Alternate names: C10orf70, MDS029, MGC14684, ZCD1, mitoNEETApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-529