Learn More
Cayman Chemical CISD1 CytosolIc DomaInhumn
SDP
Catalog No. 502948509
Encompass_Preferred
Source: Recombinant human N-terminal His-tagged CISD1 expressed in E. coli • Amino acids: 32-108 • MW: 11.3 kDa
Catalog No. 50-294-8509 Supplier Cayman Chemical Supplier No. 32034100 UG
Source: Recombinant human N-terminal His-tagged CISD1 expressed in E. coli • Amino acids: 32-108 • MW: 11.3 kDa