Learn More
Aviva Systems Biology Corp CISD2 Antibody - N-terminal region (ARP44552_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44552_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CISD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CISD2. Alternate names: ERIS, Miner1, WFS2, ZCD2, NAF-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53428