Learn More
ABclonal Technology CISD2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A141681000UL
Encompass_Preferred
The protein encoded by this gene is a zinc finger protein that localizes to the endoplasmic reticulum. The encoded protein binds an iron/sulfur cluster and may be involved in calcium homeostasis. Defects in this gene are a cause of Wolfram syndrome 2.
Catalog No. 50-001-08504