Learn More
Aviva Systems Biology Corp Cish Antibody - N-terminal region (ARP52798_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52798_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cish. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI385595, CIS1, Cis, F17, F23ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-768