Learn More
Abnova™ CISH Implementation HRP-DAB Kit
Catalog No. 89335586 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
Catalog No. For Use With (Application)
89-335-586 CISH-P
1 options
Catalog No. 89-335-586 Supplier Abnova™ Supplier No. KA5367
Add to Cart
Add to Cart
CISH Implementation HRP-DAB Kit is intended to be used in combination with digoxigenin-labeled CISH Probes
For the qualitative detection and discrimination of human pathogen viruses, e.g., HPV, EBV, CMV, and the determination of lymphocyte clonality by detecting Ig-kappa and Ig-lambda light chain mRNA in formalin-fixed, paraffin-embedded specimens by chromogenic in in situ hybridization (CISH).
Specifications
For Use With (Application) CISH-P