Learn More
Abnova™ CISH Implementation Kit 1
Catalog No. 89335584 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
Catalog No. For Use With (Application)
89-335-584 CISH-P
1 options
Catalog No. 89-335-584 Supplier Abnova™ Supplier No. KA5365
Add to Cart
Add to Cart
CISH Implementation Kit 1 is intended to be used in combination with digoxigenin-labeled CISH Probe
For the qualitative detection of genetic abbreviations, e.g., translocations, deletions, amplifications, and chromosome aneuploidies, in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
Specifications
For Use With (Application) CISH-P