Learn More
Medchemexpress LLC Cisplatin | 15663-27-1 | 99.3% | 300.05 | 1 G
SDP
Catalog No. 5000385635
Encompass_Preferred
Cisplatin (CDDP) is an antineoplastic chemotherapy agent that works by cross-linking with DNA and causing DNA damage in cancer cells. It also activates ferroptosis and induces autophagy.
  • Works by cross-linking with DNA
  • Causes DNA damage in cancer cells
  • Activates ferroptosis
  • Induces autophagy
  • Induces apoptosis in HeLa cells
  • Activates the MEK/ERK signaling pathway
  • Reduces solid tumor size and weight in melanoma-bearing mice
  • Can be used to induce acute kidney injury models
Catalog No. 50-003-85635 Supplier Medchemexpress LLC Supplier No. HY173941G
Cisplatin (CDDP) is an antineoplastic chemotherapy agent that works by cross-linking with DNA and causing DNA damage in cancer cells. It also activates ferroptosis and induces autophagy.
  • Works by cross-linking with DNA
  • Causes DNA damage in cancer cells
  • Activates ferroptosis
  • Induces autophagy
  • Induces apoptosis in HeLa cells
  • Activates the MEK/ERK signaling pathway
  • Reduces solid tumor size and weight in melanoma-bearing mice
  • Can be used to induce acute kidney injury models