Learn More
Medchemexpress LLC Cisplatin | 15663-27-1 | 99.3% | 300.05 | 50 MG
SDP
Catalog No. 5000385293
Encompass_Preferred
Cisplatin is an antineoplastic chemotherapy agent that works by cross-linking with DNA and causing DNA damage in cancer cells.
  • Activates ferroptosis
  • Induces autophagy
Catalog No. 50-003-85293 Supplier Medchemexpress LLC Supplier No. HY1739450MG
Cisplatin is an antineoplastic chemotherapy agent that works by cross-linking with DNA and causing DNA damage in cancer cells.
  • Activates ferroptosis
  • Induces autophagy