Learn More
AdipoGen Cisplatin, 15663-27-1, MFCD00011623, 250 mg
SDP
Catalog No. 501176111
Encompass_Preferred
MF: Cl2H6N2Pt, MW: 300.0, ≥98%, Appearance: Yellow solid. Potent platinum-based antineoplastic agent, Anticancer compound, Forms inter- and intrastrand DNA adducts/crosslinks, consequently blocking DNA replication and transcription and inducing cell death, Nephrotoxic when used in vivo, and potent chemoimmunotherapeutic. Stimulates the immune responses by activating macrophages and other cells of the immune system.
Catalog No. 50-117-6111 Supplier AdipoGen Supplier No. AGCR13590M250
MF: Cl2H6N2Pt, MW: 300.0, ≥98%, Appearance: Yellow solid. Potent platinum-based antineoplastic agent, Anticancer compound, Forms inter- and intrastrand DNA adducts/crosslinks, consequently blocking DNA replication and transcription and inducing cell death, Nephrotoxic when used in vivo, and potent chemoimmunotherapeutic. Stimulates the immune responses by activating macrophages and other cells of the immune system.