Learn More
Medchemexpress LLC Cisplatin (Synonyms: cis-Platinum; CDDP; cis-Diaminodichloroplatinum)
SDP
Catalog No. NC3357434
Encompass_Preferred
Cisplatin (CDDP) is an antineoplastic chemotherapy agent by cross-linking with DNA and causing DNA damage in cancer cells. Cisplatin activates ferroptosis and induces autophagy.
Catalog No. NC3357434 Supplier Medchemexpress LLC Supplier No. HY173945MG
Cisplatin (CDDP) is an antineoplastic chemotherapy agent by cross-linking with DNA and causing DNA damage in cancer cells. Cisplatin activates ferroptosis and induces autophagy.