Learn More
Abcam Cisplatin, antineoplastic agent, 100MG
Catalog No. 502706653
Encompass_Preferred
MW 300.05 Da. Potent, broad spectrum antineoplastic agent. Cytotoxic, crosslinks DNA to impair replication and transcription. Arrests cell cycle in G₂/M phase. Antitumor, cross-linking and radiation sensitizing agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6653 Supplier Abcam Supplier No. AB141398100MG
MW 300.05 Da. Potent, broad spectrum antineoplastic agent. Cytotoxic, crosslinks DNA to impair replication and transcription. Arrests cell cycle in G₂/M phase. Antitumor, cross-linking and radiation sensitizing agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement