Learn More
Enzo Life Sciences Cisplatin, (250mg), CAS Number: 15663-27-1
SDP
Supplier:  Enzo Life Sciences ALX400040M250
Encompass_Preferred
Antitumor agent. A potent platinum-based antineoplastic drug. Its mode of action involves formation of inter- and intrastrand DNA adducts which activate signaling pathways culminating in apoptosis. Platinated DNA adducts also enhance poisoning of DNA topoisomerase I. Cisplatin synergizes with a variety of anticancer agents. Tumor cells rapidly develop resistance to cisplatin and numerous agents are available to block or reverse resistance. Alternative Name: cis-Platinum(ll)diamine dichloride. Formula: Cl2H6N2Pt. MW: 300.0. CAS Number: 15663-27-1. MI: 14: 2317. Purity: ≥98% (HPLC). Appearance: Off-white to orange powder. Solubility: Soluble in dimethyl formamide (16mg/ml) or DMSO (10mg/ml); sparingly soluble in water; insoluble in 100% ethanol. Long Term Storage: Ambient
Catalog No. NC0362848