Learn More
Medchemexpress LLC Cistanoside A | 93236-42-1 | 99.6% | 800.75 | 5 MG
SDP
Catalog No. 5000428558
Encompass_Preferred
Cistanoside A is a phenylethanoid isolated from Cistanche deserticola. It reduces NO accumulation and exhibits an anti-inflammatory effect.
  • Reduces NO accumulation
  • No effect on iNOS mRNA, iNOS protein levels, or iNOS activity
  • Anti-inflammatory effect
Catalog No. 50-004-28558 Supplier Medchemexpress LLC Supplier No. HYN00235MG
Cistanoside A is a phenylethanoid isolated from Cistanche deserticola. It reduces NO accumulation and exhibits an anti-inflammatory effect.
  • Reduces NO accumulation
  • No effect on iNOS mRNA, iNOS protein levels, or iNOS activity
  • Anti-inflammatory effect