Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cistanoside A | 93236-42-1 | MFCD01725342 | 99.6% | 800.75 g/mol | C36H48O20 | 10 MG
SDP

Catalog No. 5000297029
Encompass_Preferred

Cistanoside A is a phenylethanoid glycoside isolated from Cistanche deserticola with reported nitric oxide (NO)-modulating and anti-inflammatory activity. It is provided as a high-purity research reagent suitable for in vitro and in vivo formulation studies.

  • High purity (99.6%) suitable for research applications.
  • Phenylethanoid glycoside with reported NO synthase activity.
  • Well characterized: CAS 93236-42-1; formula C36H48O20; MW 800.75 g/mol.
  • Soluble in DMSO at high concentration; multiple in vivo vehicle options documented.
  • Recommended storage -20°C; in solvent stability at -80°C for up to 6 months.
  • Supplied in a 10 mg quantity for small-scale experiments.

Catalog No. 50-002-97029 Supplier Medchemexpress LLC Supplier No. HYN002310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cistanoside A is a phenylethanoid glycoside isolated from Cistanche deserticola with reported nitric oxide (NO)-modulating and anti-inflammatory activity. It is provided as a high-purity research reagent suitable for in vitro and in vivo formulation studies.

  • High purity (99.6%) suitable for research applications.
  • Phenylethanoid glycoside with reported NO synthase activity.
  • Well characterized: CAS 93236-42-1; formula C36H48O20; MW 800.75 g/mol.
  • Soluble in DMSO at high concentration; multiple in vivo vehicle options documented.
  • Recommended storage -20°C; in solvent stability at -80°C for up to 6 months.
  • Supplied in a 10 mg quantity for small-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.