Learn More
Cayman Chemical cis-trismethoxy Resveratrol, Purity: ≥98%, CAS: 94608-23-8, 50 mg
SDP
Catalog No. 502952452
Encompass_Preferred
A potent anti-mitotic drug that is 100-fold more active than resveratrol at inhibiting the growth of human colon cancer Caco-2 cells; inhibits tubulin polymerization in a dose-dependent manner (IC50 = 4 μM) and enzymes involved in the synthesis of the polyamines, putrescine, and spermidine
Catalog No. 50-295-2452 Supplier Cayman Chemical Supplier No. 1319950 MG
A potent anti-mitotic drug that is 100-fold more active than resveratrol at inhibiting the growth of human colon cancer Caco-2 cells; inhibits tubulin polymerization in a dose-dependent manner (IC50 = 4 μM) and enzymes involved in the synthesis of the polyamines, putrescine, and spermidine