Learn More
Selleck Chemical LLC Citalopram E4953-50MG
SDP
Catalog No. 5000041578
Encompass_Preferred
Citalopram is a selective serotonin reuptake inhibitor (SSRI) that selectively inhibits the 5-HT transporter blocks the reuptake of 5-HT in the presynaptic membrane prolongs and increases the effects of 5-HT and enhances serotonin neurotransmission by potently and selectively inhibiting serotonin reuptake (SSRI)
Catalog No. 50-000-41578 Supplier Selleck Chemical LLC Supplier No. E495350MG
Citalopram is a selective serotonin reuptake inhibitor (SSRI) that selectively inhibits the 5-HT transporter blocks the reuptake of 5-HT in the presynaptic membrane prolongs and increases the effects of 5-HT and enhances serotonin neurotransmission by potently and selectively inhibiting serotonin reuptake (SSRI)