Learn More
Medchemexpress LLC Citalopram | 59729-33-8 | 99.8% | 1 ML
SDP
Catalog No. 5000233486
Encompass_Preferred
Citalopram is an orally active selective serotonin reuptake inhibitor (SSRI) provided for research use. This product is supplied as a 10 mM solution in DMSO (1 mL) and is intended for in vitro pharmacology and neuroscience studies.
  • Orally active selective serotonin reuptake inhibitor for research use.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • High purity (99.8%) suitable for biochemical and cell-based assays.
  • Molecular formula C20H21FN2O and molecular weight 324.39.
  • Suitable for in vitro pharmacology and neuroscience research applications.
Catalog No. 50-002-33486 Supplier Medchemexpress LLC Supplier No. HY12120310MM 1ML
Citalopram is an orally active selective serotonin reuptake inhibitor (SSRI) provided for research use. This product is supplied as a 10 mM solution in DMSO (1 mL) and is intended for in vitro pharmacology and neuroscience studies.
  • Orally active selective serotonin reuptake inhibitor for research use.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • High purity (99.8%) suitable for biochemical and cell-based assays.
  • Molecular formula C20H21FN2O and molecular weight 324.39.
  • Suitable for in vitro pharmacology and neuroscience research applications.