Learn More
Cayman Chemical Citalopram-d4 (hydrobromide), Purity: ≥99% deuterated forms (d1-d4), CAS: 1219803-58-3, 1 mg
SDP
Catalog No. 502986629
Encompass_Preferred
An internal standard for the quantification of citalopram by GC- or LC-MS
Catalog No. 50-298-6629 Supplier Cayman Chemical Supplier No. 221021 MG
An internal standard for the quantification of citalopram by GC- or LC-MS