Learn More
Medchemexpress LLC Citalopram (hydrobromide) | 59729-32-7 | 1 ML
SDP
Catalog No. 5000412081
Encompass_Preferred
Citalopram hydrobromide is a selective serotonin reuptake inhibitor (SSRI) with blood-brain barrier permeability. It inhibits 5-HT uptake into synaptosomes and rabbit blood platelets, demonstrating an antidepressant effect. It also exhibits concentration-dependent cytotoxicity on various neuroblastoma cell lines and human primary Schwann cells in vitro.
  • Selective serotonin reuptake inhibitor (SSRI)
  • Blood-brain barrier permeability
  • Inhibits 5-HT uptake into synaptosomes with an IC50 of 1.8 nM
  • Inhibits 5-HT uptake in rabbit blood platelets with an IC50 of 14 nM
  • Exhibits concentration-dependent cytotoxicity on various neuroblastoma cell lines and human primary Schwann cells in vitro
Catalog No. 50-004-12081 Supplier Medchemexpress LLC Supplier No. HYB128710MM/1ML
Citalopram hydrobromide is a selective serotonin reuptake inhibitor (SSRI) with blood-brain barrier permeability. It inhibits 5-HT uptake into synaptosomes and rabbit blood platelets, demonstrating an antidepressant effect. It also exhibits concentration-dependent cytotoxicity on various neuroblastoma cell lines and human primary Schwann cells in vitro.
  • Selective serotonin reuptake inhibitor (SSRI)
  • Blood-brain barrier permeability
  • Inhibits 5-HT uptake into synaptosomes with an IC50 of 1.8 nM
  • Inhibits 5-HT uptake in rabbit blood platelets with an IC50 of 14 nM
  • Exhibits concentration-dependent cytotoxicity on various neuroblastoma cell lines and human primary Schwann cells in vitro