Learn More
Medchemexpress LLC Citalopram hydrobromide | 59729-32-7 | 99.87% | 5 MG
SDP
Catalog No. 5000413209
Encompass_Preferred
Citalopram hydrobromide is a selective serotonin reuptake inhibitor (SSRI) that exhibits blood-brain barrier permeability. It inhibits 5-HT uptake into synaptosomes with an IC50 of 1.8 nM and in rabbit blood platelets with an IC50 of 14 nM, demonstrating an antidepressant effect.
  • Selective serotonin reuptake inhibitor
  • Permeates the blood-brain barrier
  • Inhibits 5-HT uptake into synaptosomes
  • Inhibits 5-HT uptake in rabbit blood platelets
  • Exhibits antidepressant effects
  • For research use only
Catalog No. 50-004-13209 Supplier Medchemexpress LLC Supplier No. HYB12875MG
Citalopram hydrobromide is a selective serotonin reuptake inhibitor (SSRI) that exhibits blood-brain barrier permeability. It inhibits 5-HT uptake into synaptosomes with an IC50 of 1.8 nM and in rabbit blood platelets with an IC50 of 14 nM, demonstrating an antidepressant effect.
  • Selective serotonin reuptake inhibitor
  • Permeates the blood-brain barrier
  • Inhibits 5-HT uptake into synaptosomes
  • Inhibits 5-HT uptake in rabbit blood platelets
  • Exhibits antidepressant effects
  • For research use only