Learn More
Medchemexpress LLC Citalopram (hydrobromide) | 59729-32-7 | 99.9% | 100 MG
SDP
Catalog No. 5000413582
Encompass_Preferred
Citalopram hydrobromide is a selective serotonin reuptake inhibitor (SSRI), with blood-brain barrier permeability. It inhibits 5-HT uptake into synaptosomes and in rabbit blood platelets, exhibiting an antidepressant effect. It also demonstrates concentration-dependent cytotoxicity on the viability of various rat and human neuroblastoma cell lines and human primary Schwann cells.
  • Selective serotonin reuptake inhibitor (SSRI)
  • Blood-brain barrier permeability
  • Inhibits 5-HT uptake
  • Antidepressant effect
Catalog No. 50-004-13582 Supplier Medchemexpress LLC Supplier No. HYB1287100MG
Citalopram hydrobromide is a selective serotonin reuptake inhibitor (SSRI), with blood-brain barrier permeability. It inhibits 5-HT uptake into synaptosomes and in rabbit blood platelets, exhibiting an antidepressant effect. It also demonstrates concentration-dependent cytotoxicity on the viability of various rat and human neuroblastoma cell lines and human primary Schwann cells.
  • Selective serotonin reuptake inhibitor (SSRI)
  • Blood-brain barrier permeability
  • Inhibits 5-HT uptake
  • Antidepressant effect