Learn More
Medchemexpress LLC Citalopram (hydrobromide) | 59729-32-7 | 99.9% | 50 MG
SDP
Catalog No. 5000412049
Encompass_Preferred
Citalopram hydrobromide is a selective serotonin reuptake inhibitor (SSRI) that can cross the blood-brain barrier. It inhibits 5-HT uptake into synaptosomes with an IC50 of 1.8 nM and inhibits 5-HT uptake in rabbit blood platelets with an IC50 of 14 nM, demonstrating an antidepressant effect.
  • It is a selective serotonin reuptake inhibitor.
  • It exhibits blood-brain barrier permeability.
  • It inhibits 5-HT uptake into synaptosomes.
  • It inhibits 5-HT uptake in rabbit blood platelets.
  • It has an antidepressant effect.
Catalog No. 50-004-12049 Supplier Medchemexpress LLC Supplier No. HYB128750MG
Citalopram hydrobromide is a selective serotonin reuptake inhibitor (SSRI) that can cross the blood-brain barrier. It inhibits 5-HT uptake into synaptosomes with an IC50 of 1.8 nM and inhibits 5-HT uptake in rabbit blood platelets with an IC50 of 14 nM, demonstrating an antidepressant effect.
  • It is a selective serotonin reuptake inhibitor.
  • It exhibits blood-brain barrier permeability.
  • It inhibits 5-HT uptake into synaptosomes.
  • It inhibits 5-HT uptake in rabbit blood platelets.
  • It has an antidepressant effect.