Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citalopram Related Compound D Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012704 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2704 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2704 Supplier MilliporeSigma Supelco Supplier No. PHR190330MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 1188264-72-3
Linear Formula C19H19 FN2 O · HBr
Synonym 1-(4-Fluorophenyl)-1-(3-methylamino)propyl)-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide; 1-(4-Fluorophenyl)-1-(3-methylaminopropyl)-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide
Type Certified Reference Material
Quantity 30 mg
Formula Weight 391.28
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.