Learn More
Selleck Chemical LLC Citarinostat (ACY-241) 5mg 1316215-12-9 HDAC-IN-2
SDP
Catalog No. 501366079
Encompass_Preferred
Citarinostat (ACY-241, HDAC-IN-2) is an orally available selective HDAC6 inhibitor with IC50 of 2.6 nM and 46 nM for HDAC6 and HDAC3, respectively. It has 13 to 18-fold selectivity towards HDAC6 in comparison to HDAC1-3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6079 Supplier Selleck Chemical LLC Supplier No. S84645MG
Citarinostat (ACY-241, HDAC-IN-2) is an orally available selective HDAC6 inhibitor with IC50 of 2.6 nM and 46 nM for HDAC6 and HDAC3, respectively. It has 13 to 18-fold selectivity towards HDAC6 in comparison to HDAC1-3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.