Learn More
Cayman Chemical CITCO PosItIve CNTRL10 m 10uL
SDP
Catalog No. 502893446
Encompass_Preferred
A USP1 inhibitor (IC50 21.1 nM) reduces the viability of H1299 cells at 500 nM induces cell cycle arrest at the S phase in MDA-MB-436 cells at 1 UM
Catalog No. 50-289-3446 Supplier Cayman Chemical Supplier No. 60068110 UL
A USP1 inhibitor (IC50 21.1 nM) reduces the viability of H1299 cells at 500 nM induces cell cycle arrest at the S phase in MDA-MB-436 cells at 1 UM