Learn More
Aviva Systems Biology Corp CITED2 Antibody - N-terminal region (ARP35789_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35789_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CITED2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CITED2. Alternate names: MRG1, P35SRJ, ASD8, VSD2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Sheep
Catalog No. 50-014-55852