Learn More
Medchemexpress LLC HY-B0739 100mg Medchemexpress, Citicoline CAS:987-78-0 Purity:>98%
SDP
Catalog No. 502082653
Encompass_Preferred
Medchemexpress, HY-B0739 100mg Citicoline CAS:987-78-0 Citicoline (Cytidine diphosphate-choline) is an intermediate in the synthesis of phosphatidylcholine, a component of cell membranes. Citicoline exerts neuroprotective effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2653 Supplier Medchemexpress LLC Supplier No. HYB0739100MG
Medchemexpress, HY-B0739 100mg Citicoline CAS:987-78-0 Citicoline (Cytidine diphosphate-choline) is an intermediate in the synthesis of phosphatidylcholine, a component of cell membranes. Citicoline exerts neuroprotective effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.