Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citicoline, 98%

Catalog No. AC457780250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457780250 25 g
AC457780010 1 g
AC457780050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC457780250 Supplier Thermo Scientific Chemicals Supplier No. 457780250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 987-78-0 | C14H26N4O11P2 | 488.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 987-78-0
Molecular Formula C14H26N4O11P2
Molecular Weight (g/mol) 488.33
MDL Number MFCD00868097
InChI Key RZZPDXZPRHQOCG-OJAKKHQRSA-N
Synonym cdp-choline, cidifos, citicoline, cytidine diphosphate choline, cdp-choline 1-, 5'-o-2-trimethylazaniumyl ethoxy phosphinato oxy phosphinato cytidine
PubChem CID 25202509
ChEBI CHEBI:58779
IUPAC Name [[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES C[N+](C)(C)CCOP([O-])(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O

Specifications

Melting Point 240.0°C to 242.0°C
Loss on Drying 5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 488.32
Percent Purity 98%
Chemical Name or Material Citicoline
EINECSNumber 213-580-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.