Learn More
Medchemexpress LLC Citicoline-d9 (sodium) | 99.8% | 1 MG
SDP
Catalog No. 5000408932
Encompass_Preferred
Citicoline-d9 (Cytidine diphosphate-choline-d9) sodium is a deuterium-labeled compound and an endogenous intermediate in the synthesis of phosphatidylcholine, a component of cell membranes. It inhibits reactive oxygen species (ROS) and apoptosis and is used in studies related to neurological diseases and hearing loss. Stable heavy isotopes have been incorporated into this drug molecule, primarily as tracers for quantitation during drug development, and deuteration has the potential to affect pharmacokinetic and metabolic profiles.
  • Used for neurological disease and hearing loss studies
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits reactive oxygen species (ROS) and apoptosis
  • Endogenous intermediate in phosphatidylcholine synthesis
  • For research use only
Catalog No. 50-004-08932 Supplier Medchemexpress LLC Supplier No. HYB0739AS1MG
Add to Cart
Add to Cart
Citicoline-d9 (Cytidine diphosphate-choline-d9) sodium is a deuterium-labeled compound and an endogenous intermediate in the synthesis of phosphatidylcholine, a component of cell membranes. It inhibits reactive oxygen species (ROS) and apoptosis and is used in studies related to neurological diseases and hearing loss. Stable heavy isotopes have been incorporated into this drug molecule, primarily as tracers for quantitation during drug development, and deuteration has the potential to affect pharmacokinetic and metabolic profiles.
  • Used for neurological disease and hearing loss studies
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits reactive oxygen species (ROS) and apoptosis
  • Endogenous intermediate in phosphatidylcholine synthesis
  • For research use only